Dinatriumsuccinat Hexahydrat ≥98%, weißes kristallines Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Soluble in water (100 mg/ml - clear, colorless solution).
Use as a TCA (Krebs cycle) supplement in cell culture applications. Sodium succinate dibasic hexahydrate can be used to study x-ray crystallography, protein structural analysis, proteomics and optimization reagents. Sodium succinate dibasic hexahydrate has been used to determine the calorimetric and diffractometric evidence for the sequential crystallization of buffer components and the consequential pH swing in frozen solutions. Sodium succinate dibasic hexahydrate has also been used to provide insights into the mechanism of Pseudomonas dacunhae aspartate ß-decarboxylase from rapid-scanning stopped-flow kinetics.
Soluble in water (100 mg/ml - clear, colorless solution).
Formel:
NaOOCCH₂CH₂COONa·6H₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149117 CAS-Nummer: 6106-21-4 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
pH (1% aq soln) | 7.7 - 8.7 |
Moisture Content | 35 - 40% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...