Pararosaniliniumchlorid ≥88% (Farbstoffgehalt), kristallines Pulver C.I. 42500
Lieferant: MP Biomedicals
Achtung
Synonyme: Basic Parafuchsin, 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride, Basic Fuchsin hydrochloride, Parafuchsin hydrochloride, Pararosaniline chloride, Basic Red 9, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride, Paramagenta hydrochloride
0215042305 0215042325
ICNA0215042305EA 34.4 EUR
ICNA0215042305 ICNA0215042325
Pararosaniliniumchlorid ≥88% (Farbstoffgehalt), kristallines Pulver C.I. 42500
Pararosaniliniumchlorid
Basic Fuchsin is a cationic triphenylmethane dye.
  • Staining of acid fast bacteria
Basic fuchsin is used for the Ziehl-Neelsen and other similar acid fast staining of the mycobacteria which cause tuberculosis, leprosy etc. and is widely used in biology to stain the nucleus.
Soluble in water (2 mg/ml); ethanol (2 mg/ml - clear red solution).
Bitte beachten Research Use Only (RUO).
Formel: C₁₉H₁₈ClN₃
Molekulargewicht: 323.82 g/mol
Schmelzpunkt: 268
MDL Number: MFCD00001657
CAS-Nummer: 569-61-9
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
UV/Vis: λmax (50% ethanol) 544 - 550 nm