L-Norvalin, weißes kristallines Pulver
Lieferant: MP Biomedicals
Synonyme:
(S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid
0210249501
0210249505
0210249510
ICNA0210249501EA
44.9
EUR
ICNA0210249501
ICNA0210249505
ICNA0210249510
L-Norvalin, weißes kristallines Pulver
L-Norvalin
Enhances NO production from activated macrophages
Soluble in water and 3N-HCl (clear, colorless solution).
Bitte beachten Research Use Only (RUO).
Formel:
C₅H₁₁NO₂ Molekulargewicht: 117.15 g/mol Siedepunkt: 228 Schmelzpunkt: 101 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064421 CAS-Nummer: 6600-40-4 Merck Index: 12,06813 |
Spezifikation Testergebnisse
FTIR | Conforms to Standard |
Optical Rotation (c=2, 6N HCl) | +23 ± 5° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...