Carboxymethylcellulose Natriumsalz, 50 - 200 cps (4% H₂O), Pulver, niedrige Viskosität
Lieferant: MP Biomedicals
Gefahr
Synonyme: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC
0210127890 0210127891
ICNA0210127890EA 93.5 EUR
ICNA0210127890 ICNA0210127891
Carboxymethylcellulose Natriumsalz, 50 - 200 cps (4% H₂O), Pulver, niedrige Viskosität
Carboxymethyl cellulose sodium salt
Carboxymethyl ether cellulose is used in drilling muds, in detergents as a soil-suspending agent, in resin emulsion paints, adhesives, printing inks, textile sizes, as protective colloid in general. As stabiliser in foods. It is used in pharmaceuticals as a suspending agent, tablet excipient, viscosity-increasing agent. Low viscosity is usually used in thin aqueous solutions.
  • Soluble in aqueous solution at 25 °C
Carboxymethyl ether cellulose sodium salt used as suspending agents. Its viscosity is both concentration and temperature dependent. As the temperaure increases, the viscosity decreases. As the concentration increases, the viscosity increases.
Soluble in aqueous solution at 25 °C
The solid should be added to the water. Stir gently or shake intermittently; do not stir constantly with a magnetic stirring bar.
Formel: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number: MFCD00081472
CAS-Nummer: 9004-32-4