Powder pillows, chromium
Lieferant: Hach
1271099
1206699
204399
204499
HACH1271099EA
54.7
EUR
HACH1271099
HACH1206699
HACH204399
HACH204499
Powder pillows, chromium
Umwelt-Prüfkits
ChromaVer 3 reagent for hexavalent chromium determination by the Diphenylcarbohydrazide Method. USEPA accepted for reporting for wastewater analyses using Hach Method 8023.