4-(Hydroxymethyl)benzonitril 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Cyanobenzyl alcohol
454650010
454650050
454650250
ACRO454650010EA
56.1
EUR
ACRO454650010
ACRO454650050
ACRO454650250
4-(Hydroxymethyl)benzonitril 97%
4-(Hydroxymethyl)benzonitril
Formel:
C₈H₇NO Schmelzpunkt: 41…42 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00870633 CAS-Nummer: 874-89-5 UN-Nummer: 3439 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Colorless to white to yellow Low melting solid |
Infrared spectrum | Conforms |
Melting point | 38 to 44 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...