Lithium aluminium di-n-butylamide 0.16 M in 1,2-Dimethoxyethan

Lieferant: Thermo Fisher Scientific
Gefahr

449191000 449195000
ACRO449191000EA 124 EUR
ACRO449191000 ACRO449195000
Lithium aluminium di-n-butylamide 0.16 M in 1,2-Dimethoxyethan
Lithium aluminium di-n-butylamide
Formel: C₃₂H₇₂AlLiN₄
Schmelzpunkt: –54 °C
Dichte: 0,86 g/cm³
Flammpunkt: –6 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00051918
CAS-Nummer: 15405-86-4
UN-Nummer: 1993
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Solution (May develop some turbidity or precipitate)
Strength 0.14 to 0.18 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR