Potassium trimethylsilanolate 2 M in THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: trimethylsiloxypotassium, potassium and trimethyl(oxido)silane, Silanol, 1,1,1-trimethyl-, potassium salt (1:1)

448461000 448468000
448461000.EA 84.2 EUR
448461000. 448468000.
Potassium trimethylsilanolate 2 M in THF, AcroSeal®
Potassium trimethylsilanolate
Formel: C₃H₉KOSi
Dichte: 0.91 g/cm³
Flammpunkt: –17 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00013421
CAS-Nummer: 10519-96-7
UN-Nummer: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to straw yellow Solution
Titration with HClO4 1.9 to 2.2 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR