2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzimidazole, 2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole
445160010
445160050
ACRO445160010EA
193
EUR
ACRO445160010
ACRO445160050
2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester 97%
2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester
Formel:
C₁₄H₁₉BN₂O₂ Schmelzpunkt: 211…212 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD13176536 CAS-Nummer: 1314216-34-6 |
Spezifikation Testergebnisse
Appearance | White to yellow Solid |
Infrared spectrum | Conforms |
Melting point | 205 to 211 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...