Fmoc-D-phenylalanin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Fmoc-D-Phe-OH, N-Fmoc-D-phenylalanine
441710010
441710050
ACRO441710010EA
21
EUR
ACRO441710010
ACRO441710050
Fmoc-D-phenylalanin 98%
Fmoc-D-phenylalanin
Formel:
C₂₄H₂₁NO₄ Molekulargewicht: 387.4 g/mol Schmelzpunkt: 184…186.5 °C Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00062955 CAS-Nummer: 86123-10-6 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Specific optical rotation | +38° to +42° (20°C, 589 nm) (C=1 IN DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...