N-TIPS indole-3-boronic acid pinacol ester 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-1-triisopropylsilanyl-1H-indole, 2-[1-(Triisopropylsilyl)-1H-indole-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

441030010 441030050
ACRO441030010EA 160 EUR
ACRO441030010 ACRO441030050
N-TIPS indole-3-boronic acid pinacol ester 97%
N-TIPS indole-3-boronic acid pinacol ester
Formel: C₂₃H₃₈BNO₂Si
Schmelzpunkt: 111…115 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16659107
CAS-Nummer: 476004-82-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white to pink Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR