Pyridoxamine dihydrochloride 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride

436250010 436250050
ACRO436250010EA 61.3 EUR
ACRO436250010 ACRO436250050
Pyridoxamine dihydrochloride 98%
Pyridoxamine dihydrochloride
Formel: C₈H₁₂N₂O₂·2HCl
Schmelzpunkt: 216…218 °C
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD00012808
CAS-Nummer: 524-36-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to light brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 to ≤102.5 % (on dry basis)
Water ≤10 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR