4-Methylaminophenolsulfat 99,0-101,5% ACS

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: p-(Methylamino)phenol sulphate, Metol, 4-Methylaminophenol sulphate, 4-(Methylamino)phenol hemisulphate, 4-Methylaminophenolsulphate, 4-(Methylamino)phenol hemisulphate salt

434441000 434445000
ACRO434441000EA 68.4 EUR
ACRO434441000 ACRO434445000
4-Methylaminophenolsulfat 99,0-101,5% ACS
4-Methylaminophenolsulfat
Formel: C₁₄H₂₀N₂O₆S
Siedepunkt: 227 °C (1013 hPa)
Schmelzpunkt: 260 °C (dec.)
Dichte: 1,434 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00013140
CAS-Nummer: 55-55-0
UN-Nummer: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder (Discolors in air)
Residue after ignition ≤0.1 %
Titration 99.0 to 101.5 %
Suitability (for determ. of phosphate) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR