(R)-(+)-1-Boc-3-methylpiperazin 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(3R)-3-Methylpiperazine, N1-BOC protected, (R)-1-Boc-3-Methylpiperazine, (R)-1-Boc-3-methyl-piperazine
432050010
432050050
ACRO432050010EA
63.5
EUR
ACRO432050010
ACRO432050050
(R)-(+)-1-Boc-3-methylpiperazin 99%
(R)-(+)-1-Boc-3-methylpiperazin
Formel:
C₁₀H₂₀N₂O₂ Schmelzpunkt: 44.4…46.6 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02683205 CAS-Nummer: 163765-44-4 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 43 to 47 °C |
GC | ≥98.5 % |
Specific optical rotation | +14° to +18° (20 °C, 589 nm) (c=1, chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...