2-Chlorothiazole-4-carboxylic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

429550010 429550050
ACRO429550010EA 52.6 EUR
ACRO429550010 ACRO429550050
2-Chlorothiazole-4-carboxylic acid 97%
2-Chlorothiazole-4-carboxylic acid
Formel: C₄H₂ClNO₂S
Schmelzpunkt: 220 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09870033
CAS-Nummer: 5198-87-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to orange Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR