Erbium(III)perchlorat 40% (w/w) in Wasser
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
428290050
428290250
ACRO428290050EA
76.3
EUR
ACRO428290050
ACRO428290250
Erbium(III)perchlorat 40% (w/w) in Wasser
Erbium(III) perchlorate
Formel:
Cl₃ErO₁₂ Dichte: 1.44 g/cm³ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00016073 CAS-Nummer: 14017-55-1 UN-Nummer: 3211 ADR: 5.1,II |
Spezifikation Testergebnisse
Appearance | Clear pink to purple Liquid |
Titration Complexometric | 14 to 15 % (Er) |
Thiosulphate (S2O3) | ≤5000 ppm |
Sulfite (as SO2) | ≤2000 ppm |
Total nitrogen (as N) | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...