Bromessigsäure-N-hydroxysuccinimid-Ester 90%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
427780010
427780050
427782500
ACRO427780010EA
831
EUR
ACRO427780010
ACRO427780050
ACRO427782500
Bromessigsäure-N-hydroxysuccinimid-Ester 90%
Bromessigsäure-N-hydroxysuccinimid-Ester
Formel:
C₆H₆BrNO₄ Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00058571 CAS-Nummer: 42014-51-7 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Elemental Analysis | ≥88 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...