N-Acetyl-D-mannosamine 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Cyclic N-Acetyl-D-mannosamine, N-((2R,3S,4R,5S,6R)-2,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide
427530010
427530050
ACRO427530010EA
0
EUR
ACRO427530010
ACRO427530050
N-Acetyl-D-mannosamine 98%
N-Acetyl-D-mannosamine
Formel:
C₈H₁₅NO₆ Molekulargewicht: 221.21 g/mol (anhy) Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00149493 CAS-Nummer: 7772-94-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤11 % |
HPLC | ≥97.5 % (ELSD) |
Specific optical rotation |
+9.5° to +10.5° (20°C, 589 nm) (c=4 in H2O) reading after |
Iron (Fe) | ≤0.2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...