Amberlite® FPC3500 Ionenaustauscherharz
Lieferant: Thermo Fisher Scientific
389992500
389990010
ACRO389992500EA
37.9
EUR
ACRO389992500
ACRO389990010
Amberlite® FPC3500 Ionenaustauscherharz
Amberlite® FPC3500
Lagerungstemperatur:
Raumtemperatur |
MDL Number:
MFCD08460528 CAS-Nummer: 950182-46-4 |
Spezifikation Testergebnisse
Appearance | White to light yellow Beads |
Particle size | ≤5.0 % (> 1.180 mm) |
Harmonic mean size | 0.450 to 0.650 mm |
Uniformity coefficient | ≤2.00 |
Moisture holding capacity | 60 to 70 % (H) |
Total volume capacity | ≥2.60 eq/l (H) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...