N-ɑ-Fmoc-N-Trityl-L-histidine 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-ɑ-Fmoc-tele-trityl-L-histidine, Fmoc-His(Trt)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-trityl-1H-imidazol-4-yl)propanoic acid, N-ɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-tele-(triphenylmethyl)-L-histidine, N-Fmoc-1-trityl-L-histidine

360330050 360330250
ACRO360330050EA 25.2 EUR
ACRO360330050 ACRO360330250
N-ɑ-Fmoc-N-Trityl-L-histidine 98%
N-alpha-Fmoc-N-Trityl-L-histidine
Formel: C₄₀H₃₃N₃O₄
Molekulargewicht: 619.72 g/mol
Schmelzpunkt: 133…135 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00043332
CAS-Nummer: 109425-51-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1.0 %
HPLC ≥97.5 %
Appearance of solution (0.5mmol/ml DMF) clear, colourless
Specific optical rotation +80.0° to +89.0° (20°C, 589 nm) (c=1, chloroform) 0.2% D-enantiomer
Residual solvents ≤5 % (2-Propanol, THF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR