Methyl mercaptan sodium salt 95%, rein
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Sodium methanethiolate, Sodium thiomethoxide, Sodium methylsulphanide, Methanethiol sodium salt
305710050
305710010
305710250
305711000
ACRO305710050EA
106
EUR
ACRO305710050
ACRO305710010
ACRO305710250
ACRO305711000
Methyl mercaptan sodium salt 95%, rein
Methyl mercaptan sodium salt
Formel:
CH₃NaS Molekulargewicht: 70.08 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00174316 CAS-Nummer: 5188-07-8 UN-Nummer: 2921 ADR: 8 I |
Spezifikation Testergebnisse
Appearance | White to light beige Powder or granular powder |
Titration Iodimetric | 94 to 106 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...