Amberlite® IRA-410 (Cl) Ionenaustauscherharz

Lieferant: Thermo Fisher Scientific

Synonyme: Amberlite® IRA-410 Cl type

202291000 202295000
ACRO202291000EA 23.3 EUR
ACRO202291000 ACRO202295000
Amberlite® IRA-410 (Cl) Ionenaustauscherharz
Amberlite® IRA-410 (Cl)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00132710
CAS-Nummer: 9002-26-0

Bestellen

Spezifikation Testergebnisse

Appearance Transparent light yellow to light orange Beads
Particle size ≤5.0 % (>1.180 mm)
Harmonic mean size 0.600 to 0.750 mm
Uniformity coefficient ≤1.60
Moisture holding capacity 45.0 to 51.0 % (Cl-)
Total volume capacity ≥1.25 eq/l (Cl-)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR