Bromoacetic anhydride 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
426700050
426700250
ACRO426700050EA
144
EUR
ACRO426700050
ACRO426700250
Bromoacetic anhydride 95%
Bromoacetic anhydride
Appearance: White to brown Solid
Formel:
C₄H₄Br₂O₃ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00045041 CAS-Nummer: 13094-51-4 UN-Nummer: 3265 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to brown Solid |
Infrared spectrum | Conforms |
Melting point | 34 to 42 °C |
Titration Argentometric | ≥94 % (ex Br) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...