6-Carboxyfluorescein 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM
425801000
425805000
ACRO425801000EA
127
EUR
ACRO425801000
ACRO425805000
6-Carboxyfluorescein 96%
6-Carboxyfluorescein
Appearance: Yellow to orange Powder
Formel:
C₂₁H₁₂O₇ Schmelzpunkt: >300 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00036873 CAS-Nummer: 3301-79-9 |
Spezifikation Testergebnisse
Appearance | Yellow to orange Powder |
Infrared spectrum | Conforms |
HPLC | ≥95 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...