N-Boc-DL-valin 98%

Lieferant: Thermo Fisher Scientific

Synonyme: BOC-DL-Valine, Boc-DL-Val-OH, 2-((tert-Butoxycarbonyl)amino)-3-methylbutanoic acid

425780100 425780500
ACRO425780100EA 70.5 EUR
ACRO425780100 ACRO425780500
N-Boc-DL-valin 98%
N-Boc-DL-valin
Formel: C₁₀H₁₉NO₄
Schmelzpunkt: 112…114 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00069973
CAS-Nummer: 54895-12-4

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
NMR consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR