2-Chlorophenylacetyl chloride 97%

Lieferant: Thermo Fisher Scientific
Gefahr

395080050 395080250
ACRO395080050EA 37.3 EUR
ACRO395080050 ACRO395080250
2-Chlorophenylacetyl chloride 97%
2-Chlorophenylacetyl chloride

Appearance: Clear yellow Liquid

Formel: C₈H₆Cl₂O
Dichte: 1,31 g/cm³
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00130223
CAS-Nummer: 51512-09-5
UN-Nummer: 3265
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5500 (20°C, 589 nm)
Titration RCOCl ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR