5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
AICAR, 5-Amino-1-(β-D-ribofuranosyl)-1H-imidazole-4-carboxamide, Acadesine
398920050
398920250
398921000
ACRO398920050EA
56.7
EUR
ACRO398920050
ACRO398920250
ACRO398921000
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside 98%
5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside
Appearance: White to tan Crystalline powder
Formel:
C₉H₁₄N₄O₅ Schmelzpunkt: 214…216 °C Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00869751 CAS-Nummer: 2627-69-2 |
Spezifikation Testergebnisse
Appearance | White to tan Crystalline powder |
Infrared spectrum | Conforms |
TLC | ≥97.5 % |
Specific optical rotation | -62.0° to -66.0° (20°C, 589 nm) (c=1 H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...