Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate 94%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bis(norbornadiene)rhodium(I) tetrafluoroborate, Bis(norbornadiene)rhodium (I) tetrafluoroborate
382551000
382555000
ACRO382551000EA
72.9
EUR
ACRO382551000
ACRO382555000
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate 94%
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate
Appearance: Red Powder
Formel:
C₁₄H₈BF₄Rh Schmelzpunkt: 209…211 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00671775 CAS-Nummer: 36620-11-8 UN-Nummer: 1325 ADR: 4.1,II |
Spezifikation Testergebnisse
Appearance | Red Powder |
Infrared spectrum | Conforms |
Elemental Analysis | ≥92 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...