Nocodazole 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl (6-(thiophene-2-carbonyl)-1H-benzo[d]imidazol-2-yl)carbamate
358240100
358240500
ACRO358240100EA
109
EUR
ACRO358240100
ACRO358240500
Nocodazole 98%
Nocodazole
Appearance: White to orange-brown Powder
Formel:
C₁₄H₁₁N₃O₃S Schmelzpunkt: 300…305 °C Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00005588 CAS-Nummer: 31430-18-9 |
Spezifikation Testergebnisse
Appearance | White to orange-brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...