2-Aminoethyl methacrylate hydrochloride 90% stabilisiert

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Aminoethyl 2-methylprop-2-enoate hydrochloride

357810050 357810250
ACRO357810050EA 63.1 EUR
ACRO357810050 ACRO357810250
2-Aminoethyl methacrylate hydrochloride 90% stabilisiert
2-Aminoethyl methacrylate hydrochloride

Appearance: White to light green to light brown Crystalline powder

Formel: C₆H₁₂ClNO₂
Schmelzpunkt: 116…118 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00078260
CAS-Nummer: 2420-94-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light green to light brown Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥88 %
Stabilizer 500 to 1000 ppm phenothiazine


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR