(S)-(+)-2-Aminobuttersäure 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-Abu-OH, L-alpha-Amino-N-Butyric acid, L-2-Aminobutyric acid, (S)-2-Aminobutanoic acid
215380010
215380050
ACRO215380010EA
40.3
EUR
ACRO215380010
ACRO215380050
(S)-(+)-2-Aminobuttersäure 98%
(S)-(+)-2-Aminobuttersäure
Formel:
C₄H₉NO₂ Schmelzpunkt: >300 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064415 CAS-Nummer: 1492-24-6 |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % (K.F.) |
Sulfated ash | ≤0.1 % |
Specific optical rotation | +20.2° to +23° (20°C, 589 nm) (c=2, 5 N HCl) |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...