Gallocyanin

Lieferant: Thermo Fisher Scientific
Achtung

154430100 154430250
ACRO154430100EA 26.7 EUR
ACRO154430100 ACRO154430250
Gallocyanin
Gallocyanine

Appearance: Dark green to black Crystalline powder

Formel: C₁₅H₁₃ClN₂O₅
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011926
CAS-Nummer: 1562-85-2

Bestellen

Spezifikation Testergebnisse

Appearance Dark green to black Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR