Acid red 27 85%, rein

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Amaranth, Sodium 3-hydroxy-4-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-2,7-disulphonate, Bordeaux S, Food Red No. 2, Azo Rubine S, Wool Red 40 F, Naphthol Red

153030250 153031000 153035000
ACRO153030250EA 60.1 EUR
ACRO153030250 ACRO153031000 ACRO153035000
Acid red 27 85%, rein
Acid red 27

CAS No.: 915-67-3

Formel: C₂₀H₁₄N₂O₁₀S₃
Molekulargewicht: 604.46 g/mol
Schmelzpunkt: 218…220 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00004076
CAS-Nummer: 915-67-3

Bestellen

Spezifikation Testergebnisse

Appearance Dark red-brown to brown Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥80 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR