p-Toluenesulphinic acid sodium salt hydrate 98+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Sodium-p-toluenesulphinate hydrate, 4-Methylbenzenesulphinic acid sodium salt hydrate
139011000
139015000
ACRO139011000EA
45.6
EUR
ACRO139011000
ACRO139015000
p-Toluenesulphinic acid sodium salt hydrate 98+%
p-Toluenesulphinic acid sodium salt hydrate
CAS No.: 207801-20-5
Formel:
C₇H₇NaO₂S .xH₂O Schmelzpunkt: >300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149640 CAS-Nummer: 207801-20-5 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder or flakes |
Infrared spectrum | Conforms |
Water | ≤5 % (K.F.) |
Titration with HClO4 | ≥98.0 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...