2-Carboxyphenyl salicylate 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Salicyloyloxybenzoic Acid, O-Salicyloylsalicylic Acid, Salicylic Acid 2-Carboxyphenyl Ester, Salsalate

133400250 133401000
ACRO133400250EA 52.5 EUR
ACRO133400250 ACRO133401000
2-Carboxyphenyl salicylate 98%
2-Carboxyphenyl salicylate

CAS No.: 552-94-3

Formel: C₁₄H₁₀O₅
Schmelzpunkt: 139…151 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00020252
CAS-Nummer: 552-94-3

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %
NMR (1H-NMR) Consistent with structure
NMR (13C-NMR) Consistent with structure
NMR (31P-NMR) Consistent with structure
Elemental Analysis 76.05 to 82.40 % C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR