3,4,9,10-Perylenetetracarboxylic dianhydride 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Anthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone, Pigment Red 224, PTCDA

130081000 130080250
ACRO130081000EA 104 EUR
ACRO130081000 ACRO130080250
3,4,9,10-Perylenetetracarboxylic dianhydride 98%
3,4,9,10-Perylenetetracarboxylic dianhydride

CAS No.: 128-69-8

Formel: C₂₄H₈O₆
Schmelzpunkt: >350 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006916
CAS-Nummer: 128-69-8

Bestellen

Spezifikation Testergebnisse

Appearance Dark red Powder
Infrared spectrum Conforms
Assay ≥97.5 %
Water ≤1 % (K.F.)
Stabilizer 150 to 400 ppm (BHT)
Peroxides ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR