ɑ-Amino-γ-butyrolactone hydrobromide 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: DL-Homoserine lactone hydrobromide

103290100 103290025 103290500
ACRO103290100EA 148 EUR
ACRO103290100 ACRO103290025 ACRO103290500
ɑ-Amino-γ-butyrolactone hydrobromide 99%
alpha-Amino-gamma-butyrolactone hydrobromide

CAS No.: 6305-38-0

Formel: C₄H₇NO₂
Schmelzpunkt: 221 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00012723
CAS-Nummer: 6305-38-0

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Dye content ≥65 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR