N-Acetyl-L-tyrosine ethyl ester monohydrate 99%

Lieferant: Thermo Fisher Scientific

102750010 102750100 102750500
ACRO102750010EA 26.7 EUR
ACRO102750010 ACRO102750100 ACRO102750500
N-Acetyl-L-tyrosine ethyl ester monohydrate 99%
N-Acetyl-L-tyrosine ethyl ester monohydrate
Formel: C₁₃H₁₉NO₅
Schmelzpunkt: 80…81 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149093
CAS-Nummer: 36546-50-6

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Water 3.5 to 8.0 % (K.F.)
HPLC ≥98.5 %
Appearance of solution (5% soln. in EtOH) clear, colorless
Specific optical rotation +22° to +24° (24°C, 589 nm) (c=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR