ɑ-Methylene-γ-butyrolactone 95% stabilisiert
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Methylenedihydro-2(3H)-furanone, 3-Methylenedihydrofuran-2(3H)-one
414260010
414260050
ACRO414260010EA
44.1
EUR
ACRO414260010
ACRO414260050
ɑ-Methylene-γ-butyrolactone 95% stabilisiert
alpha-Methylene-gamma-butyrolactone
Appearance: Clear colorless to light yellow to light pink Liquid
Formel:
C₅H₆O₂ Molekulargewicht: 98.10 g/mol Siedepunkt: 88…92 °C (12 mmHg) Dichte: 1.12 g/cm³ Flammpunkt: 37 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00005407 CAS-Nummer: 547-65-9 UN-Nummer: 1993 ADR: 3 III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow to light pink Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4710 to 1.4750 (20°C, 589 nm) |
GC | ≥94 % |
Stabilizer | 2,6-di-tert.-Butyl-p-cresol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...