Ethyl 2-Bromoisovalerate 95%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Bromoisovaleric Acid Ethyl Ester, 2-Bromo-3-methylbutyric Acid Ethyl Ester, Ethyl 2-bromo-3-methylbutanoate, Ethyl 2-Bromo-3-methylbutyrate

409790100 409790500
ACRO409790100EA 64.6 EUR
ACRO409790100 ACRO409790500
Ethyl 2-Bromoisovalerate 95%
Ethyl-2-bromoisovalerate

CAS No.: 609-12-1

Formel: C₇H₁₃BrO₂
Siedepunkt: 77 °C (12 mmHg)
Flammpunkt: 65 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00026855
CAS-Nummer: 609-12-1

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4480 to 1.4510 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR