Barium chloranilate trihydrate
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Chloranilic acid barium salt, Barium 3,6-Dichloro-1,4-dioxo-3,6-cyclohexadiene-2,5-diolate
401660250
401661000
ACRO401660250EA
151
EUR
ACRO401660250
ACRO401661000
Barium chloranilate trihydrate
Barium chloranilate trihydrate
Appearance: Brown Powder
Formel:
C₆H₂Cl₂O₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150175 CAS-Nummer: 32458-20-1 UN-Nummer: 1564 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Brown Powder |
Infrared spectrum | Conforms |
Suitability | (for Sulfate determination) Passes test |
Titration with NaOH | 94 to 108 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...