N-Pivaloyl-o-toluidine 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-(2-Chlorophenyl)-2,2-dimethylpropanamide, N-(o-tolyl)pivalamide
346730050
346730250
ACRO346730050EA
97.3
EUR
ACRO346730050
ACRO346730250
N-Pivaloyl-o-toluidine 99%
N-Pivaloyl-o-toluidine
Appearance: White Crystalline powder
Formel:
C₁₂H₁₇NO Schmelzpunkt: 111…115 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00075432 CAS-Nummer: 61495-04-3 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 111 to 115 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...