DL-2-Benzylserine 97%

Lieferant: Thermo Fisher Scientific
Achtung

339780010 339780100
ACRO339780010EA 57.5 EUR
ACRO339780010 ACRO339780100
DL-2-Benzylserine 97%
DL-2-Benzylserine

Appearance: Beige to light brown Crystalline powder or flakes

Formel: C₁₀H₁₃NO₃
Schmelzpunkt: 245…263 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01863305
CAS-Nummer: 4740-47-0

Bestellen

Spezifikation Testergebnisse

Appearance Beige to light brown Crystalline powder or flakes
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR