Creatinphosphat Dinatriumsalz Tetrahydrat 99%

Lieferant: Thermo Fisher Scientific

Synonyme: Sodium creatine phosphate dibasic tetrahydrate, Sodium N-carbamimidoyl-N-methylglycinate dihydrogen phosphate tetrahydrate, Phosphocreatine disodium salt tetrahydrate

337340100 337340010 337340020
ACRO337340100EA 251 EUR
ACRO337340100 ACRO337340010 ACRO337340020
Creatinphosphat Dinatriumsalz Tetrahydrat 99%
Creatinphosphat Dinatriumsalz Tetrahydrat
Formel: C₄H₈N₃Na₂O₅P·4H₂O
Molekulargewicht: 327.14 g/mol (255.07 anhy)
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00150192
CAS-Nummer: 71519-72-7

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Water 21 to 25 % (K.F.)
Solubility (10 % in water) Clear colorless solution
Titration with HClO4 98.5 to 101.5 %
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR