1,3,6,8-Pyrenetetrasulphonic acidtetrasodium salt 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
1,3,6,8-Pyrenetetrasulphonic acid tetrasodium salt
325560010
325560050
ACRO325560010EA
60.8
EUR
ACRO325560010
ACRO325560050
1,3,6,8-Pyrenetetrasulphonic acidtetrasodium salt 95%
1,3,6,8-Pyrenetetrasulphonic acidtetrasodium salt
Appearance: Green Crystalline powder
Formel:
C₁₆H₆Na₄O₁₂S₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00042029 CAS-Nummer: 59572-10-0 |
Spezifikation Testergebnisse
Appearance | White to yellow to green Crystalline powder |
Infrared spectrum | Conforms |
UV spectrophotometry | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...