Nonafluorovaleric acid 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
309290050
309290250
ACRO309290050EA
83
EUR
ACRO309290050
ACRO309290250
Nonafluorovaleric acid 97%
Nonafluorovaleric acid
Formel:
C₅HF₉O₂ Siedepunkt: 140 °C Dichte: 1,71 g/cm³ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00040211 CAS-Nummer: 2706-90-3 UN-Nummer: 3265 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Clear light yellow to light brown Liquid |
Infrared spectrum | Conforms |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...