Methylselenocysteine 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Se-methylseleno-L-cysteine, (R)-2-Amino-3-(methylselanyl)propanoic acid, (R)-2-Amino-3-(methylseleno)propionic acid, Se-(Methyl)seleno-L-cysteine, H-3-(Me-Se)-Ala-OH

300642500 300640010 300640500
ACRO300642500EA 323 EUR
ACRO300642500 ACRO300640010 ACRO300640500
Methylselenocysteine 98%
Methylselenocysteine

Appearance: White to light yellow Powder

Formel: C₄H₉NO₂Se
Molekulargewicht: 182.092 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00800565
CAS-Nummer: 26046-90-2
UN-Nummer: 3283
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder
Infrared spectrum Conforms
Specific optical rotation -13° to -15° (20°C, 589 nm) (c=1, water)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR