Triisopropanolamine cyclic borate 98+%
Lieferant: Thermo Fisher Scientific
Synonyme:
3,7,10-Trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo[3.3.3]undecane, Triisopropanolamine Borate, Triisopropanolamine Cyclic Borate
269241000
269245000
ACRO269241000EA
179
EUR
ACRO269241000
ACRO269245000
Triisopropanolamine cyclic borate 98+%
Triisopropanolamine cyclic borate
CAS No.: 101-00-8
Formel:
C₉H₁₈BNO₃ Schmelzpunkt: 150…157 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03791298 CAS-Nummer: 101-00-8 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline needles and/or chunks |
Infrared spectrum | Conforms |
Melting point | 150 to 157 °C |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...