Erioglaucine disodium salt, rein
Lieferant: Thermo Fisher Scientific
Synonyme:
FD and C Blue No. 1, Brilliant Blue FCF, Erioglaucin A, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulphonate, Alphazurine FG, Erioglaucine sodium salt, CI 42090, Acid blue 9, Food Blue No.1
229730050
229730250
ACRO229730050EA
35.7
EUR
ACRO229730050
ACRO229730250
Erioglaucine disodium salt, rein
Erioglaucine disodium salt
Appearance: Dark purple Crystalline powder
Formel:
C₃₇H₃₆N₂O₉S₃ Molekulargewicht: 792.84 g/mol Schmelzpunkt: 283 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012141 CAS-Nummer: 3844-45-9 |
Spezifikation Testergebnisse
Appearance | Dark purple Crystalline powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Loss on drying | ≤10 % (105°C) |
UV Lambda max | 625 to 630 nm |
Absorption Ratio P-15/P+15 | 1.10 to 1.25 |
Specific absorbance A (1%/1cm) | ≥1000 (at lambda max.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...