Povidone iodine

Lieferant: Thermo Fisher Scientific

Synonyme: Polyvinylpyrrolidone-iodine complex

229471000 229470025 229475000
ACRO229471000EA 97.6 EUR
ACRO229471000 ACRO229470025 ACRO229475000
Povidone iodine
Povidone iodine

CAS No.: 25655-41-8

Formel: C₆H₉I₂NO
Molekulargewicht: 364.953 g/mol
Schmelzpunkt: 300 °C (Zers.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00084483
CAS-Nummer: 25655-41-8

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to red-brown Powder
Infrared spectrum Conforms
Assay 9.0 to 12.0 % (available iodine) (on dried substance)
Loss on drying ≤8 % (5 g, 105°C)
Sulfated ash ≤0.025 % (2 g)
Iodide (I) ≤6.6 % (on dried substance)
Nitrogen (N) 9.5 to 11.5 % (on dried substance)
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR